Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MPAYHSSLMD PDTKLIGNMA LLPIRSQFKG PAPRETKDTD IVDEAIYYFK ANVFFKNYEI KNEADRTLIY ITLYISEC
This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been conserved through evolution and is implicated in the control of actin polymerization in cells. Alternatively spliced transcript variants have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin related protein 2/3 complex subunit 3, 21kDa; actin related protein 2/3 complex, subunit 3, 21kDa; Actin-related protein 2/3 complex subunit 3; Arp2/3 complex 21 kDa subunit; ARP2/3 protein complex subunit p21; p21-ARC
Gene Aliases: ARC21; ARPC3; p21-Arc
UniProt ID: (Human) O15145
Entrez Gene ID: (Human) 10094
Molecular Function: actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support