Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TATLRPYLS AVRATLQAAL CLENFSSQVV ERHNKPEVEV RSSKELLLQP VTISRNEKEK VLIEGSINSV RVSIAVKQAD EIEKILCHKF MRFTMMRAEN FFILRRKPVE GYDISFLITN FHTEQMYKHK LVDFVIHFME EIDKEISEMK LSVNARARIV AEEFLKNF (1-168 aa encoded by BC012596)
This gene encodes one of seven subunits of the human Arp2/3 protein complex. This complex controls actin polymerization in cells and has been conserved throughout eukaryotic evolution. This gene encodes the p20 subunit, which is necessary for actin nucleation and high-affinity binding to F-actin. Alternative splicing results in multiple transcript variants. Naturally occurring read-through transcription exists between this gene and the downstream tubulin tyrosine ligase-like family, member 3, which results in the production of a fusion protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin related protein 2/3 complex subunit 4, 20kDa; actin related protein 2/3 complex, subunit 4, 20kDa; Actin-related protein 2/3 complex subunit 4; Arp2/3 complex 20 kDa subunit; arp2/3 protein complex subunit p20; MGC13544; p20-ARC
Gene Aliases: 20kDa; 5330419I20Rik; AI327076; ARC20; ARPC4; P20-ARC
UniProt ID: (Human) P59998, (Mouse) P59999
Entrez Gene ID: (Human) 10093, (Mouse) 68089, (Rat) 297518
Molecular Function:
actin or actin-binding cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support