Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSRIYHDGAL RNKAVQSVRL PGAWDPAAHQ GGNGVLLEGE LIDVSRHSIL DTHGRKERYY VLYIRPSHIH RRKF
Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.
ARPIN gene ontology annotations related to this gene include negative regulation of actin nucleation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arp2/3 inhibition protein; Arpin; UPF0552 protein C15orf38
Gene Aliases: ARPIN; C15orf38
UniProt ID: (Human) Q7Z6K5
Entrez Gene ID: (Human) 348110
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support