Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MIKAGVAGCR EPQFIYPNII GRAKGQSRAA QGGLELCVGD QAQDWRSSLF ISYPVERGLI TSCEDMEIMW KHIYDYNLKL KPCDGPVLIT EPALNPLANR QQITEMFFEH LGVPAFYMSI QAVLALFAAG FTTGLVLNSG AGVTQSVPIF EGYCLPHGVQ QLDLAGLDLT NYLMVLMKNH GIMLLSASDR KIVEDIKESF CYVAMNYEEE MAKKPDCL
ACTRT3 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: actin related protein M1; actin, gamma 1 propeptide; Actin-related protein M1; Actin-related protein T3; ARP-T3
Gene Aliases: ACTRT3; ARP-T3; ARPM1; RGD1561457
UniProt ID: (Human) Q9BYD9
Entrez Gene ID: (Human) 84517, (Rabbit) 100343735, (Rat) 365763
Molecular Function: actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support