Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LVFLLADDL GWNDVGFHGS RIRTPHLDAL AAGGVLLDNY YTQPLCTPSR SQLLTGRYQI RTGLQHQIIW PCQPSCVPLD EKLLPQLLKE AGYTTHMVGK WHLGMYRKEC LPTRRGFDTY FGYLLGSEDY YSHERCTLID ALNVTRCALD FRDGEEVATG YKNMYSTNIF TKRAIALITN HPPEKPLFLY LALQSVHEPL QVPEEYLKPY DFIQDKNRHH YAGMVSLMDE AVGNVTAALK SSGLWNNTVF IFSTDNGGQT LAGGNNWPLR GRKWSLWEGG VRGVGFVASP LLKQKGVKNR ELIHISDWLP TLVKLARGHT NGTKPLDGFD VWKTISEGSP SPRIELLHNI DPNFVDSSPY WPECSLLL (47-413 aa encoded by BC029051 )
Arylsulfatase B encoded by this gene belongs to the sulfatase family. The arylsulfatase B homodimer hydrolyzes sulfate groups of N-Acetyl-D-galactosamine, chondriotin sulfate, and dermatan sulfate. The protein is targetted to the lysozyme. Mucopolysaccharidosis type VI is an autosomal recessive lysosomal storage disorder resulting from a deficiency of arylsulfatase B. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arylsulfatase B; Arylsulfatase B (MPS VI); arylsulfatase B complex; arylsulfatase B regulation; arylsulfatase B structural; arylsulfatase B temporal regulation; ASB; G4S; N-acetylgalactosamine-4-sulfatase
Gene Aliases: 1110007C02Rik; AI480648; ARSB; As-1; As-1r; As-1s; As-1t; As1; As1-r; As1-s; As1-t; ASB; Asr-1; Ast-1; G4S; MPS6
UniProt ID: (Human) P15848, (Rat) P50430, (Mouse) P50429
Entrez Gene ID: (Human) 411, (Rat) 25227, (Mouse) 11881
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support