Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LKISQAPQVS ISRSKSYREN GAPFVPPPPA LDELGGPGGV THPEERLGVA GGPGSAPAAG GGTGTEDDEE ELLEDEEDED EEEELLEDDE EELLEDDARA LLKEPR
Arx contains two conserved domains, a C-peptide (or aristaless domain) and the prd-like class homeobox domain. It is a member of the group-II aristaless-related protein family whose members are expressed primarily in the central and/or peripheral nervous system. This protein is thought to be involved in CNS development. Mutations in the Arx gene cause X-linked mental retardation and epilepsy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aristaless-related homeobox; aristaless-related homeobox, X-linked; cancer/testis antigen 121; Homeobox protein ARX
Gene Aliases: ARX; CT121; EIEE1; ISSX; MRX29; MRX32; MRX33; MRX36; MRX38; MRX43; MRX54; MRX76; MRX87; MRXS1; PRTS
UniProt ID: (Human) Q96QS3
Entrez Gene ID: (Human) 170302
Molecular Function: homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support