Transform your discoveries into market-ready therapies

Biotech solutions

Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • ASAH1 Antibodies

            Proteintech

            ASAH1 Polyclonal Antibody

            11 References
            View all (14) ASAH1 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite ASAH1 Polyclonal Antibody

            • Antibody Testing Data (11)
            Application
            ASAH1 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 11

            ASAH1 Antibody (11274-1-AP) in ICC/IF

            Immunofluorescent analysis of SH-SY5Y cells using 11274-1-AP (ASAH1 antibody) at dilution of 1:25 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L).
            View Product

            ASAH1 Antibody in Immunocytochemistry (ICC/IF)
            ASAH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ASAH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ASAH1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ASAH1 Antibody in Western Blot (WB)
            ASAH1 Antibody in Western Blot (WB)
            ASAH1 Antibody in Western Blot (WB)
            ASAH1 Antibody in Western Blot (WB)
            ASAH1 Antibody in Western Blot (WB)
            ASAH1 Antibody in Western Blot (WB)
            ASAH1 Antibody in Immunoprecipitation (IP)

            Product Details

            11274-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:1,000
            View 9 publications 9 publications

            Immunohistochemistry (IHC)

            -
            View 1 publication 1 publication

            Immunohistochemistry (Paraffin) (IHC (P))

            1:20-1:200
            -

            Immunocytochemistry (ICC/IF)

            1:10-1:100
            View 1 publication 1 publication

            Immunoprecipitation (IP)

            0.5-4.0 µg
            -
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Published species

            Human, Mouse

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            ASAH1 Fusion Protein Ag1799 (1-389 aa encoded by BC016828)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.15 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            Immunogen sequence: MNCCIGLGE KARGSHRASY PSLSALFTEA SILGFGSFAV KAQWTEDCRK STYPPSGPTV FPAVIRYRGA VPWYTINLDL PPYKRWHELM LDKAPVPGLL GNFPGPFEEE MKGIAAVTDI PLGEIISFNI FYELFTVCTS IVAEDKKGHL IHGRNMDFGV FLGWNINNDT WVITEQLKPL TVNLDFQRNN KTVFKASSFA GYVGMLTGFK PGLFSLTLNE RFSINGGYLG ILEWILGKKD AMWIGFLTRT VLENSTSYEE AKNLLTKTKI LAPAYFILGG NQSGEGCVIT RDRKESLDVY ELDAKQGRWY VVQTNYDRWK HPFFLDDRRT PAKMCLNRTS QENISFETMY DVLSTKPVLN KLTVYTTLID VTKGQFETYL RDCPDPCIGW (1-389 aa encoded by BC016828)

            Target Information

            ASAH1 encodes a member of the acid ceramidase family of proteins. Alternative splicing results in multiple transcript variants, at least one of which encodes a preproprotein that is proteolytically processed. The processing of this preproprotein generates alpha and beta subunits that heterodimerize to form the mature lysosomal enzyme, which catalyzes the degradation of ceramide into sphingosine and free fatty acid. This enzyme is overexpressed in multiple human cancers and may play a role in cancer progression. Mutations in this gene have been linked to the lysosomal storage disorder, Farber lipogranulomatosis, and a neuromuscular disorder, spinal muscular atrophy with progressive myoclonic epilepsy. The related pathways include Innate Immune System and Sphingolipid metabolism. Diseases associated with ASAH1 include Farber Lipogranulomatosis and Spinal Muscular Atrophy With Progressive Myoclonic Epilepsy.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: AC; ACDase; acid CDase; Acid ceramidase; Acylsphingosine deacylase; Glycosylceramide deacylase; N-acylethanolamine hydrolase ASAH1; N-acylsphingosine amidohydrolase; N-acylsphingosine amidohydrolase (acid ceramidase) 1; N-acylsphingosine amidohydrolase 1; PHP32; Putative 32 kDa heart protein

            View more View less

            Gene Aliases: 2310081N20Rik; AC; ACDase; ASAH; ASAH1; HSD-33; HSD33; PHP; PHP32; SMAPME

            View more View less

            UniProt ID: (Human) Q13510, (Mouse) Q9WV54, (Rat) Q6P7S1

            View more View less

            Entrez Gene ID: (Human) 427, (Mouse) 11886, (Rat) 84431

            View more View less

            Function(s)

            catalytic activity ceramidase activity hydrolase activity molecular_function cysteine protease

            Process(es)

            ceramide metabolic process glycosphingolipid metabolic process lipid metabolic process response to organic substance lung development
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-nsvbw:80/100.66.79.248:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0