Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MNCCIGLGEK ARGSHRASYP SLSALFTEAS ILGFGSFAVK AQWTEDCRKS TYPPSGPTVF PAVIRYRGAV PWYTINLDLP PYKRWHELML DKAPVPGLLG NFPGPFEEEM KGIAAVTDIP LGEIISFNIF YELFTVCTSI VAEDKKGHLI HGRNMDFGVF LGWNINNDTW VITEQLKPLT VNLDFQRNNK TVFKASSFAG YVGMLTGFKP GLFSLTLNER FSINGGYLGI LEWILGKKDA MWIGFLTRTV LENSTSYEEA KNLLTKTKIL APAYFILGGN QSGEGCVITR DRKESLDVYE LDAKQGRWYV VQTNYDRWKH PFFLDDRRTP AKMCLNRTSQ ENISFETMYD VLSTKPVLNK LTVYTTLIDV TKGQFETYLR DCPDPCIGW
ASAH1 encodes a member of the acid ceramidase family of proteins. Alternative splicing results in multiple transcript variants, at least one of which encodes a preproprotein that is proteolytically processed. The processing of this preproprotein generates alpha and beta subunits that heterodimerize to form the mature lysosomal enzyme, which catalyzes the degradation of ceramide into sphingosine and free fatty acid. This enzyme is overexpressed in multiple human cancers and may play a role in cancer progression. Mutations in this gene have been linked to the lysosomal storage disorder, Farber lipogranulomatosis, and a neuromuscular disorder, spinal muscular atrophy with progressive myoclonic epilepsy. The related pathways include Innate Immune System and Sphingolipid metabolism. Diseases associated with ASAH1 include Farber Lipogranulomatosis and Spinal Muscular Atrophy With Progressive Myoclonic Epilepsy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AC; ACDase; acid CDase; Acid ceramidase; Acylsphingosine deacylase; Glycosylceramide deacylase; N-acylethanolamine hydrolase ASAH1; N-acylsphingosine amidohydrolase; N-acylsphingosine amidohydrolase (acid ceramidase) 1; N-acylsphingosine amidohydrolase 1; PHP32; Putative 32 kDa heart protein
Gene Aliases: AC; ACDase; ASAH; ASAH1; HSD-33; HSD33; PHP; PHP32; SMAPME
UniProt ID: (Human) Q13510, (Rat) Q6P7S1
Entrez Gene ID: (Human) 427, (Rabbit) 100338209, (Pig) 100512932, (Rat) 84431
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support