Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: IYSIFQLAF AAIFLGFYVA YKKCHGKNKA EIPESKENGT EPESSFYKAN GGFQPDEK (292-348 aa encoded by BC130523)
This gene encodes a sodium/bile acid cotransporter. This transporter is the primary mechanism for uptake of intestinal bile acids by apical cells in the distal ileum. Bile acids are the catabolic product of cholesterol metabolism, so this protein is also critical for cholesterol homeostasis. Mutations in this gene cause primary bile acid malabsorption (PBAM); mutations in this gene may also be associated with other diseases of the liver and intestines, such as familial hypertriglyceridemia (FHTG).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apical sodium-dependent bile acid transporter; ASBT; IBAT; ileal apical sodium-dependent bile acid transporter; Ileal Na(+)/bile acid cotransporter; Ileal sodium-dependent bile acid transporter; Ileal sodium/bile acid cotransporter; Na(+)-dependent ileal bile acid transporter; Sodium/taurocholate cotransporting polypeptide, ileal; solute carrier family 10 (sodium/bile acid cotransporter family), member 2; solute carrier family 10 (sodium/bile acid cotransporter), member 2; Solute carrier family 10 member 2
Gene Aliases: ASBT; IBAT; ISBT; NTCP2; PBAM; SLC10A2
UniProt ID: (Human) Q12908, (Mouse) P70172
Entrez Gene ID: (Human) 6555, (Mouse) 20494
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support