Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEVLRPQLI RIDGRNYRKN PVQEQTYQHE EDEEDFYQGS MECADEPCDA YEVEQTPQGF RSTLRAPSLL YKHIVGKRGD TRKKIEMETK TSISIPKPGQ DGEIVITGQH RNGVISARTR IDVLLDTFRR KQPFTHFLAF FLNEVEVQEG FLRFQEEVLA KCSMDHGVDS SIFQNPKKLH LTIGMLVLLS EEEIQQTCEM LQQCKEEFIN DISGGKPLEV EMAGIEYMND DPGMVDVLYA KVHMKDGSNR LQELVDRVLE RFQASGLIVK EWNSVKLHAT VMNTLFRKDP NAEGRYNLYT AEGKYIFKER ESFDGRNILK LFENFYFGSL KLNSIHISQR FTVDSFGNYA SCGQID (1-355 aa encoded by B C012291)
This gene encodes a subunit of the activating signal cointegrator 1 (ASC-1) complex. The ASC-1 complex is a transcriptional coactivator that plays an important role in gene transactivation by multiple transcription factors including activating protein 1 (AP-1), nuclear factor kappa-B (NF-kB) and serum response factor (SRF). The encoded protein contains an N-terminal KH-type RNA-binding motif which is required for AP-1 transactivation by the ASC-1 complex. Mutations in this gene are associated with Barrett esophagus and esophageal adenocarcinoma. Alternatively spliced transcripts encoding multiple isoforms have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activating signal cointegrator 1 complex subunit 1; ASC-1 complex subunit P50; Trip4 complex subunit p50
Gene Aliases: 1810015P09Rik; AI550520; Asc-1; ASC1p50; ASCC1; CGI-18; p50; SMABF2
UniProt ID: (Human) Q8N9N2, (Mouse) Q9D8Z1
Entrez Gene ID: (Human) 51008, (Rat) 294512, (Mouse) 69090
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support