Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: CHRTRETLP PFPTRLLELI EGFMLSKGLL TACKLKVFDL LKDEAPQKAA DIASKVDASA CGMERLLDIC AAMGLLEKTE QGYSNTETAN VYLASDGEYS LHGFIMHNND LTWNLFTYLE FAIREGTNQH HRALGKKAED LFQDAYYQSP ETRLRFMRAM HGMTKLTACQ VATAFNLSRF SSACDVGGCT GALARELARE YPRMQVTVFD LPDIIELAAH FQPPGPQAVQ IHFAAGDFFR DPLPSAELYV LCRILHDWPD DKVHKLLSKV AESCKPGAGL LLVETLLDEE KRVAQRALMQ SLNMLVQTEG KERSLGEYQC LLELHGFHQV QVVHLGGVLD AILATKVAP (274-621 aa encoded by BC002508)
The protein encoded by this gene has an N-terminus that is similar to the multicopy associated filamentation (maf) protein of Bacillus subtilis and to orfE of Escherichia coli, while the C-terminus is similar to N-acetylserotonin O-methyltransferase. This gene is located in the pseudoautosomal region 1 (PAR1) of X and Y chromosomes. Three transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acetylserotonin N-methyltransferase-like; Probable bifunctional dTTP/UTP pyrophosphatase/methyltransferase protein
Gene Aliases: ASMTL; ASMTLX; ASMTLY; ASTML
UniProt ID: (Human) O95671
Entrez Gene ID: (Human) 8623
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support