Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 11
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Specifically catalyzes the NAD or NADP-dependent dehydrogenation of L-aspartate to iminoaspartate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aspartate dehydrogenase domain-containing protein; putative L-aspartate dehydrogenase
Gene Aliases: 0610012D14Rik; ASPDH; RGD1310111
UniProt ID: (Human) A6ND91, (Rat) Q5I0J9, (Mouse) Q9DCQ2
Entrez Gene ID: (Human) 554235, (Rat) 292875, (Mouse) 68352
Molecular Function:
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support