Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EFYNRFKGR NDLMEYAKQH GIPIPVTPKN PWSMDENLMH ISYEAGILEN PKNQAPPGLY TKTQDPAKAP NTPDILEIEF KKGVPVKVTN VKDGTTHQTS LELFMYLNEV AGKHGVGRID IVENRFIGMK SRGIYETPAG TILYHAHLDI EAFTMDREVR KIKQGLGLKF AELVYTGFWH SPECEFVRHC IAKSQERVEG KVQVSVLKGQ VYILGRESPL SLYNEELVSM NVQGDYEPTD ATGFININSL RLKEYHRLQS KVTAK (149-412 aa encoded by BC009243)
The protein encoded by this gene catalyzes the penultimate step of the arginine biosynthetic pathway. There are approximately 10 to 14 copies of this gene including the pseudogenes scattered across the human genome, among which the one located on chromosome 9 appears to be the only functional gene for argininosuccinate synthetase. Mutations in the chromosome 9 copy of ASS cause citrullinemia. Two transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Argininosuccinate synthase; argininosuccinate synthetase 1; Ass1; Citrulline--aspartate ligase; citrulline-aspartate ligase; RP11-618A20.2
Gene Aliases: ASS; ASS1; CTLN1
UniProt ID: (Human) P00966
Entrez Gene ID: (Pig) 414411, (Human) 445
Molecular Function:
ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support