Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GRSVLLEQP RKSGSKVISH MLSSHGGEIF LHVLSSSRSI LEDPPSISEG CGGRVTDYRI TDFGEFMREN RLTPFLDPRY KIDGSLEVPL ERAKDQLEKH TRYWPMIISQ TTIFNMQAVV PLASVIVKES LTEEDVLNCQ KTIYNLVDME RKNDPLPIST VGTRGKGPKR DEQYRIMWNE LETLVRAHIN NSEKHQRVLE CLMACRSKPP EEEERKKRGR KREDKEDKSE KAVKDYEQEK SWQDSERLKG ILERGKEELA EAEIIKDSPD SPEPPNKKPL VEMDETPQVE KSKGPVSLLS LWSNRINTAN SRKHQEFAGR LNSVNNRAEL YQHLKEENGM ETTKNGKASR Q (357-706 aa encoded by BC003081)
Crucial regulator of the mitotic cell cycle and development. At prophase, required for dynein anchoring to the nuclear envelope important for proper centrosome-nucleus coupling. At G2/M phase, may be required for proper spindle formation and execution of cytokinesis. Probable component of the Integrator (INT) complex, a complex involved in the small nuclear RNAs (snRNA) U1 and U2 transcription and in their 3'-box-dependent processing (PubMed:23904267). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: asunder spermatogenesis regulator; asunder, spermatogenesis regulator homolog (Drosphila); Cell cycle regulator Mat89Bb homolog; Germ cell tumor 1; Integrator complex subunit 13; Protein asunder homolog; Sarcoma antigen NY-SAR-95; spermatogenesis associated 30
Gene Aliases: ASUN; C12orf11; GCT1; INTS13; Mat89Bb; NET48; SPATA30
UniProt ID: (Human) Q9NVM9
Entrez Gene ID: (Human) 55726
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support