Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAASALRGLP VAGGGESSES EDDGWEIGYL DRTSQKLKRL LPIEEKKEKF KKAMTIGDVS LVQELLDSGI SVDSNFQYGW TPLMYAASVA NAELVRVLLD RGANASFEKD KQSILITACS AHGSEEQILK CVELLLSRNA DPNVACRRLM TPIMYAARDG HTQVVALLVA HGAEVNTQDE NGYTALTWAA RQGHKNIVLK LLELGANKML QTKDGKMPSE IAKRNKHHEI FNLLSFTLNP LEGKLQQLTK EDTICKILTT DSDREKDHIF SSYTAFGDLE VFLHGLGLEH MTDLLKERDI TLRHLLTMRE DEFTKNGITS KDQQKILAAL KELQVEEIQF GELSEETKLE ISGDEFLNFL LKLNKQCGHL ITAVQNVITE LPVNSQKVFY KSY
Plays a central role during spermatogenesis by repressing transposable elements and preventing their mobilization, which is essential for the germline integrity. Acts via the piRNA metabolic process, which mediates the repression of transposable elements during meiosis by forming complexes composed of piRNAs and Piwi proteins and governs the methylation and subsequent repression of transposons. Its association with pi-bodies suggests a participation in the primary piRNAs metabolic process. Required prior to the pachytene stage to facilitate the production of multiple types of piRNAs, including those associated with repeats involved in the regulation of retrotransposons. May act by mediating protein-protein interactions during germ cell maturation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin repeat, SAM and basic leucine zipper domain-containing protein 1; ankyrin-like 1; Ankyrin-like protein 1; ASZ1; Germ cell-specific ankyrin, SAM and basic leucine zipper domain-containing protein
Gene Aliases: ALP1; ANKL1; ASZ1; C7orf7; CT1.19; GASZ; Orf3
UniProt ID: (Human) Q8WWH4
Entrez Gene ID: (Human) 136991
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support