Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibodies Inc
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
For maximum recovery of product, centrifuge the vial prior to removing the cap.
This gene encodes a protein that localizes to clathrin-coated pits, where it acetylates alpha tubulin on lysine 40. This process may be important in microtubule growth, for instance during chemotaxis and the formation of cilium. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acetyltransferase mec-17 homolog; acetyltransferase mec-17 homolog {ECO:0000255|HAMAP-Rule:MF_03130}; Alpha-TAT; alpha-TAT {ECO:0000255|HAMAP-Rule:MF_03130}; alpha-TAT1; alpha-TAT1 {ECO:0000255|HAMAP-Rule:MF_03130}; Alpha-tubulin N-acetyltransferase 1; alpha-tubulin N-acetyltransferase 1 {ECO:0000255|HAMAP-Rule:MF_03130}; Novel DUF738 containing protein (2610110G12Rik); TAT; TAT {ECO:0000255|HAMAP-Rule:MF_03130}
Gene Aliases: 0610011P08Rik; 2610008K08Rik; 2610110G12Rik; 3110080J08Rik; alpha-TAT; alpha-TAT1; ATAT1; C6orf134; MEC17; Nbla00487; RGD1303066; TAT
UniProt ID: (Human) Q5SQI0, (Mouse) Q8K341, (Rat) Q6MG11
Entrez Gene ID: (Human) 79969, (Mouse) 73242, (Rat) 361789
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support