Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: TTEATLRMEN VDVKEEWQDE DLPRPLPEET GVELLGSPVE DTSSPPNTLN FNGAHRKRKT LVAPEI
This gene encodes a neuron-restricted protein that contains a CRAL-TRIO motif common to proteins that bind small lipophilic molecules. Mutations in this gene are associated with cerebellar ataxia, Cayman type.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ataxia cayman type protein; ataxia cerebellar Cayman type; BNIP-2-homolgy; BNIP-2-homology; BNIP-H; Cayman ataxia; Caytaxin
Gene Aliases: ATCAY; BNIP-H; CLAC; KIAA1872
UniProt ID: (Human) Q86WG3
Entrez Gene ID: (Human) 85300
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support