Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: NTATVVATT QVPSGNPQPT ISLQPLPVIL HVPVAVSSQP QLLQSHPGTL VTNQPSGNVE FISVQSPPTV SGLTKNPVSL PSLPNPTKPN NVPSVPSPSI QRNPTASAAP LGTTLAVQAV PTAHSIVQAT RTSLPTVGPS GLYSPSTNRG PIQMKIPISA FSTSSAAEQN SNTTPRIENQ TNKTIDASVS KKAADSTSQC GKATGSDSSG VIDLTMDDEE SGASQDPKKL NHTPVSTMSS SQPVSRPLQP IQPAPPLQPS GVPTSGPSQT TIHLLPTAPT TVNVTHRPVT QVTTRLPVPR APANHQVVYT TLPAPPAQAP LRGTVMQAPA VRQVNPQNSK RFFLYMAPRY M (756-1106 aa encoded by BC063855)
ATF7IP is a multifunctional nuclear protein that associates with heterochromatin. It can act as a transcriptional coactivator or corepressor depending upon its binding partners.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activating transcription factor 7-interacting protein 1; ATF-interacting protein; ATF-IP; ATF7-interacting protein; ATFa-associated Modulator; FLJ10688; hAM; mAM; MBD1-containing chromatin-associated factor 1; P621
Gene Aliases: 2610204M12Rik; AM; ATF-IP; ATF7IP; MCAF; MCAF1; p621
UniProt ID: (Human) Q6VMQ6, (Mouse) Q7TT18
Entrez Gene ID: (Human) 55729, (Mouse) 54343, (Rat) 312800
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support