Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MTSREHQVS LCNCVPLLRR LLCDAPWRKA RPLHALSRYF RSRVSPSKMA EEPQSVLQLP TSIAAGGEGL TDVSPETTTP EPPSSAAVSP GTEEPAGDTK KKIDILLKAV GDTPIMKTKK WAVERTRTIQ GLIDFIKKFL KLVASEQLFI YVNQSFAPSP DQEVGTLYEC FGSDGKLVLH YCKSQAWG (1-187 aa encoded by BC012266)
ATG12, a member of the autophagy protein family, forms a conjugate with ATG5; this conjugate has a ubiquitin-protein ligase (E3)-like activity for protein lipidation in autophagy. This conjugate also associates with innate immune response proteins such as RIG-I and VISA (also known as IPS-1), inhibiting type I interferon production and permitting viral replication in host cells. ATG12 has also been shown to interact with ATG10 in human embryonic kidney cells in the presence of ATG7. At least two isoforms of ATG12 are known to exist. Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: APG 12; Apg12 (autophagy, yeast) homolog; APG12-like; ATG12 autophagy related 12 homolog; autophagy-related 12; Autophagy-related protein 12; OTTHUMP00000159129; Ubiquitin-like protein ATG12
Gene Aliases: 4931423H11Rik; A330058M13Rik; APG12; APG12L; ATG12; Atg12l; FBR93; HAPG12
UniProt ID: (Human) O94817, (Rat) Q2TBJ5, (Mouse) Q9CQY1
Entrez Gene ID: (Human) 9140, (Rat) 361321, (Mouse) 67526
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support