Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: DLHGIYEDGG KPPVPCLRVS KALDPKSTGR KYFLPQVVVT VNPQSSSTQW EVAPEKGEEL ELSVESPCEL REPEPSPFSS KRTMYETEEM VIPGDPEEMR T; Positive Samples: U-87MG, HeLa; Cellular Location: Lipid droplet, Peripheral membrane protein, Preautophagosomal structure membrane
Involved in autophagosome assembly, regulating the size of nascent autophagosomes (PubMed:28561066). Also regulates lipid droplets morphology and distribution within the cell (PubMed:22219374, PubMed:28561066). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATG2 autophagy related 2 homolog A; Autophagy-related protein 2 homolog A
Gene Aliases: ATG2A; KIAA0404
UniProt ID: (Human) Q2TAZ0
Entrez Gene ID: (Human) 23130
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support