Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • ATG4A Antibodies

            Osenses

            ATG4A Polyclonal Antibody

            1 Reference
            View all (18) ATG4A antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # OSA00034W-150UL
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            434.00
            150 µL
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite ATG4A Polyclonal Antibody

            Product Details

            OSA00034W-150UL

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:200-1:2,000
            -

            Immunohistochemistry (IHC)

            1:200-1:2,000
            -

            Miscellaneous PubMed (Misc)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Mouse

            Host/Isotype

            Rabbit / Ig

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            A synthetic peptide from mouse cysteine protease ATG4A (autophagin-2) conjugated to an immunogenic carrier protein has been used as the antigen

            Conjugate

            Unconjugated

            Form

            Lyophilized

            Concentration

            Conc. Not Determined

            Storage buffer

            whole serum

            Contains

            no preservative

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            Product Specific Information

            Reconstitute in 150 µL of sterile water. Centrifuge to remove any insoluble material.

            Specificity of this antibody: ATG4A.

            Target Information

            Autophagy is the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some malignant tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed. This gene encodes a member of the autophagin protein family. The encoded protein is also designated as a member of the C-54 family of cysteine proteases. Transcript variants that encode distinct isoforms have been identified.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: APG4 (ATG4) autophagy-related homolog A; AUT-like 2 cysteine endopeptidase; AUT-like 2, cysteine endopeptidase; autophagin 2; Autophagin-2; autophagy 4 homolog A; autophagy-related 4A; Autophagy-related cysteine endopeptidase 2; Autophagy-related protein 4 homolog A; cysteine protease APG4A; Cysteine protease ATG4A; MGC107179

            View more View less

            Gene Aliases: AI627006; Apg4a; Atg4a; Atg4al; Autl2; AV169859

            View more View less

            UniProt ID: (Mouse) Q8C9S8

            View more View less

            Entrez Gene ID: (Mouse) 666468

            View more View less

            Function(s)

            cysteine-type endopeptidase activity cysteine-type peptidase activity cysteine protease

            Process(es)

            autophagosome assembly mitophagy C-terminal protein lipidation proteolysis protein targeting to membrane cellular response to nitrogen starvation protein processing nucleophagy protein delipidation
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-k5nmm:80/100.66.74.94:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0