Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Cysteine protease ATG4b is an enzyme which is coded by ATG4b gene. Autophagy is a critical cellular homeostatic process that controls the turnover of damaged organelles and protein. ATG4b is a deubiquitin-like protease that plays dual roles in the core machinery of autophagy. ATG4b, cleaves the C-terminal amino acid of ATG8 family proteins MAP1LC3, GABARAPL1, GABARAPL2 and GABARAP, to reveal a C-terminal glycine. Exposure of the glycine at the C-terminus is essential for ATG8 proteins conjugation to phosphatidylethanolamine (PE) and insertion to membranes, which is necessary for autophagy. ATG4b is mainly expressed in the skeletal muscle, brain, heart, liver and pancreas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: APG4 (ATG4) autophagy-related homolog B; APG4 autophagy 4 homolog B; ATG4 autophagy related 4 homolog B; AUT-like 1 cysteine; AUT-like 1 cysteine endopeptidase; AUT-like 1, cysteine endopeptidase; autophagin 1; Autophagin-1; Autophagin-2B; autophagy related 4B, cysteine peptidase; autophagy-related 4B; Autophagy-related cysteine endopeptidase 1; Autophagy-related cysteine endopeptidase 2B; Autophagy-related protein 4 homolog B; bAut2B; Cysteine protease ATG4B; cysteine protease involved in autophagy APG4-B; hAPG4B; HsAPG4B; MmAPG4B
Gene Aliases: 2510009N07Rik; APG4B; ATG4B; Atg4bl; AUT2B; Aut2b2; AUTL1; AW048066; KIAA0943
UniProt ID: (Bovine) Q6PZ03, (Human) Q9Y4P1, (Mouse) Q8BGE6
Entrez Gene ID: (Bovine) 408002, (Human) 23192, (Mouse) 66615
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support