Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
ATG4C is involved in autophagy, the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some malignant tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed. This gene encodes a member of the autophagin protein family. The encoded protein is also designated as a member of the C-54 family of cysteine proteases. Alternate transcriptional splice variants, encoding the same protein, have been characterized. Cysteine protease required for autophagy, which cleaves the C-terminal part of either MAP1LC3, GABARAPL2 or GABARAP, allowing the liberation of form I. A subpopulation of form I is subsequently converted to a smaller form (form II). Form II, with a revealed C-terminal glycine, is considered to be the phosphatidylethanolamine (PE)-conjugated form, and has the capacity for the binding to autophagosomes. Highly expressed in skeletal muscle, heart, liver and testis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: APG4 autophagy 4 homolog C; ATG4 autophagy related 4 homolog C; AUT-like 1, cysteine endopeptidase; AUT-like 3 cysteine endopeptidase; Autophagin-3; autophagy related 4C, cysteine peptidase; Autophagy-related cysteine endopeptidase 3; Autophagy-related protein 4 homolog C; cysteine peptidase; Cysteine protease ATG4C; FLJ14867; HsAPG4C
Gene Aliases: APG4-C; APG4C; ATG4C; AUTL1; AUTL3
UniProt ID: (Human) Q96DT6
Entrez Gene ID: (Human) 84938
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support