Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In Western Blot, a band is seen ~56 kDa representing the ATG5-ATG12 complex, the molecular weight of human ATG5 is ~33 kDa.
Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended.
Suggested positive control: mouse wildtype ES cell lysate.
ATG5 (Autophagy Related 5) is an important element for autophagy and may play an important role in the apoptotic process. ATG5 is also involved in other cellular processes that include mitochondrial quality control after oxidative damage, negative regulation of the innate anti-viral immune response, lymphocyte development and proliferation, MHC II antigen presentation, and adipocyte differentiation. Following conjugation to ATG12, the conjugate participates in the formation of autophagosome. ATG5 contributes to autophagic cell death by interacting with Fas-associated protein with death domain (FADD). The ATG5-ATG12 conjugate forms a cup-shaped isolation membrane that then detaches from the membrane immediately before or after autophagosome formation is completed. APG5 may play a role in the apoptotic process, possibly within the modified cytoskeleton. Further, APG5 expression is a relatively late event in the apoptotic process, occurring downstream of caspase activity. The APG5-APG12 conjugate also associates with innate immune response proteins such as RIG-I and VISA (also known as IPS-1), inhibiting type I interferon production and permitting viral replication in host cells. Diseases associated with ATG5 dysfunction include spinocerebellar ataxia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: APG 5; APG5 autophagy 5 like (APG5L); APG5-like; Apoptosis-specific protein; Apoptosis-specific protein (ASP); ATG5 autophagy related 5 homolog; Atg5-PA; autophagy 5-like protein; Autophagy protein 5; autophagy related 5-like protein; autophagy-related 5; autophagy-specific gene 5; CG1643-PA; LD34980p
Gene Aliases: 2010107M05Rik; 3110067M24Rik; APG5; APG5-LIKE; APG5L; ASP; Atg-5; ATG5; Atg5l; AW319544; C88337; CG1643; DmAtg5; Dmel\CG1643; Dmel_CG1643; hAPG5; Paddy; XELAEV_18027227mg; zgc:100934
UniProt ID: (Pig) Q3MQ04, (Human) Q9H1Y0, (Bovine) Q3MQ24, (Fruit fly) Q9W3R7, (Mouse) Q99J83
Entrez Gene ID: (Pig) 100739102, (Human) 9474, (Bovine) 532686, (Fruit fly) 31666, (Guinea pig) 100735170, (Mouse) 11793, (Rat) 365601, (Zebrafish) 494180, (Xenopus) 443756
Molecular Function: membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support