Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene was identified based on homology to Pichia pastoris GSA7 and Saccharomyces cerevisiae APG7. In the yeast, the protein appears to be required for fusion of peroxisomal and vacuolar membranes. The protein shows homology to the ATP-binding and catalytic sites of the E1 ubiquitin activating enzymes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: APG7-like; ATG12-activating enzyme E1 ATG7; ATG7 autophagy related 7 homolog; autophagy related 7 homolog; autophagy-related 7; Autophagy-related protein 7; DKFZp434N0735; hAGP7; Ubiquitin-activating enzyme E1-like protein; Ubiquitin-like modifier-activating enzyme ATG7
Gene Aliases: 1810013K23Rik; Agp7; APG7-LIKE; APG7L; ATG7; Atg7l; Gm21553; GSA7
UniProt ID: (Human) O95352, (Rat) Q641Y5, (Mouse) Q9D906
Entrez Gene ID: (Human) 10533, (Pig) 100462749, (Rat) 312647, (Mouse) 74244
Molecular Function: ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support