Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Keep as concentrated solution.
Predicted reactivity: Mouse (96%), Rat (98%), Rhesus Monkey (99%), Bovine (98%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a GTPase and a Golgi body transmembrane protein. The encoded protein can form a homotetramer and has been shown to interact with spastin and with mitogen-activated protein kinase kinase kinase kinase 4. This protein may be involved in axonal maintenance as evidenced by the fact that defects in this gene are a cause of spastic paraplegia type 3. Three transcript variants encoding two different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATL1 alpha; ATL1 beta; ATL1 gamma; ATL1-delta; Atlastin-1; Bcl11B; BCL11B/TRDC fusion; Brain-specific GTP-binding protein; Ctip 2; CTIP-2; CTIP2; GBP-3; GTP-binding protein 3; Guanine nucleotide-binding protein 3; guanylate-binding protein 3; hGBP3; hRIT1 alpha; Rit 1; Rit1; Spastic paraplegia 3 protein A; Spastic paraplegia 3A homolog; ZNF856B
Gene Aliases: 4930435M24Rik; AD-FSP; Adfsp; ATL1; atlastin1; FSP1; GBP3; HSN1D; SPG3; SPG3A
UniProt ID: (Human) Q8WXF7, (Mouse) Q8BH66
Entrez Gene ID: (Human) 51062, (Mouse) 73991
Molecular Function: heterotrimeric G-protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support