Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Recommended positive controls: NT2D1, IMR32, U87-MG.
Predicted reactivity: Mouse (96%), Rat (96%), Zebrafish (82%), Chimpanzee (100%), Bovine (96%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ATL3 is GTPase tethering membranes through formation of trans-homooligomer and mediating homotypic fusion of endoplasmic reticulum membranes. ATL3 is function in endoplasmic reticulum tubular network biogenesis.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: atlastin 3; Atlastin-3
Gene Aliases: ATL3; HSN1F; RGD1309871
UniProt ID: (Human) Q6DD88, (Rat) Q0ZHH6
Entrez Gene ID: (Human) 25923, (Rat) 309187
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support