Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The Drosophila atonal gene produces a protein with basic helix loop helix (bHLH) domains that plays an essential role in the development of the Drosophila nervous system. Mammalian atonal homolog 1 (MATH-1) is a helix-loop-helix (HLH) transcription factor that is structurally homologous to the product of the Drosophila proneural gene atonal. MATH-1, so known as Atoh1, Ath1 or HATH-1, is a 351 amino acid protein with an atonal-related basic HLH domain. In mice, expression of MATH-1 takes place by embryonic day 9. 5 and initially localizes to the cranial ganglions and the dorsal part of the central nervous system. Prominent expression of MATH-1 is in the dorsal part of the central nervous system but becomes restricted to the external granular layer of the cerebellum by day 18 and is undetectable in the adult nervous system. It is suggested that MATH-1 may play a role in the differentiation of subsets of neural cells by activating E box-dependent transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Atonal bHLH transcription factor 1; atonal homolog 1; atonal homolog bHLH transcription factor 1; bHLHa14; Class A basic helix-loop-helix protein 14; H MATH-1; hATH1; Helix-loop-helix protein hATH-1; Helix-loop-helix protein mATH-1; mATH1; Protein atonal homolog 1; Transcription factor ATOH1
Gene Aliases: ATH1; ATOH1; BHLHA14; HATH1; MATH-1; Math1; RGD1565171
UniProt ID: (Human) Q92858, (Mouse) P48985
Entrez Gene ID: (Human) 474, (Mouse) 11921, (Rat) 500156
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support