Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: PTQLQLARQL TRKSPRRCSA PKETFAQGRL PKDSGTEHSS GRTVKTSVPL SQPSWHTQQP VCSLEASGEP STVDMSMPVR EHTLLE
Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 55%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aminophospholipid translocase VA; ATPase class V type 10A; ATPase type IV, phospholipid transporting (P-type); ATPase, class V, type 10A; ATPase, Class V, type 10C; P4-ATPase flippase complex alpha subunit ATP10A; Phospholipid-transporting ATPase VA
Gene Aliases: ATP10A; ATP10C; ATPVA; ATPVC; KIAA0566
UniProt ID: (Human) O60312
Entrez Gene ID: (Human) 57194
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support