Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SATGEIYLFC KGADSSIFPR VIEGKVDQIR ARVERNAVEG LRTLCVAYKR LIQEEYEGIC KLLQAAKVAL QDREKKLAEA YEQIEKDLTL LG
Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
ATP11A is a widely expressed integral membrane ATPase. ATP11A is probably phosphorylated in its intermediate state and likely drives the transport of ions such as calcium or other molecules across membranes. While the exact molecule ATP11A transports is unknown, increased expression of ATP11A mRNA has been observed in murine leukemia cells made resistant to anti-cancer drugs such as farnesyltransferase inhibitors (FTIs). Furthermore, overexpression of ATP11A provided protection against the FTI SCH66336 while knockdown of ATP11A via siRNA made cells more sensitive to this drug. Other reports suggest that elevated levels of ATP11A mRNA may also be a predictive marker of metastasis in colorectal cancer patients.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase class VI type 11A; ATPase IS; ATPase, class VI, type 11A; P4-ATPase flippase complex alpha subunit ATP11A; phospholipid-translocating ATPase; Phospholipid-transporting ATPase IH; potential phospholipid-transporting ATPase IH; RP11-120K24.1
Gene Aliases: ATP11A; ATPIH; ATPIS; KIAA1021
UniProt ID: (Human) P98196
Entrez Gene ID: (Human) 23250
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support