Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Adding 0.2 mL of distilled water will yield a concentration of 500 µg/mL.
The function of ATP11C remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase class VI type 11C; ATPase IQ; ATPase, class VI, type 11C; P4-ATPase flippase complex alpha subunit ATP11C; Phospholipid-transporting ATPase IG; probable phospholipid-transporting ATPase IG
Gene Aliases: ATP11C; ATPIG; ATPIQ
UniProt ID: (Human) Q8NB49
Entrez Gene ID: (Human) 286410
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support