Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
MA3928A647 detects sodium/potassium ATPase alpha from human, mouse, bovine, sheep, canine, chicken and porcine tissues. This antibody detects the alpha 3 isoform in rat only.
The MA3928A647 antigen is purified sheep kidney alpha sodium/potassium ATPase. This antibody recognizes an epitope between amino acid residues 646 and 652 of the sheep kidney alpha sodium/potassium ATPase.
The protein encoded by this gene belongs to the family of P-type cation transport ATPases, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. This enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The catalytic subunit of Na+/K+ -ATPase is encoded by multiple genes. This gene encodes an alpha 1 subunit. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (Na+ + K+)-ATPase; (Na+, K+)-ATPase alpha-subunit; ATP1B; ATPase; ATPase, Na+/K+ transporting, alpha 1 polypeptide; ATPase, Na+K+ transporting, alpha 1; MGC1798; Na(+)/K(+) ATPase alpha-1 subunit; Na+ ,K+ -ATPase alpha subunit; Na+ /K+ ATPase alpha 1 subunit; Na+, K+-ATPase beta-subunit precursor; Na+/K+ ATPase 1; Na+/K+ ATPase Alpha; Na+/K+ transporting alpha 1 polypeptide; Na, K-ATPase, alpha-A catalytic polypeptide; Na,K-ATPase alpha-1 subunit; Na,K-ATPase catalytic subunit alpha-A protein; RP4-655J12.1; Sodium Potassium ATPase; sodium pump 1; Sodium pump subunit alpha-1; sodium-potassium ATPase catalytic subunit alpha-1; sodium-potassium-ATPase, alpha 1 polypeptide; sodium/potassium-transporting ATPase alpha-1 chain; Sodium/potassium-transporting ATPase subunit alpha-1; unnamed protein product
Gene Aliases: ATP1A1; Atpa-1; BC010319; Nkaa1b
UniProt ID: (Sheep) P04074, (Human) P05023, (Pig) P05024, (Bovine) Q08DA1, (Chicken) P09572, (Rat) P06685, (Mouse) Q8VDN2
Entrez Gene ID: (Dog) 403992, (Sheep) 443381, (Human) 476, (Pig) 397481, (Bovine) 282144, (Chicken) 396530, (Rat) 24211, (Mouse) 11928
Molecular Function: primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support