Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • ATP1A3 Antibodies

            Invitrogen

            ATP1A3 Monoclonal Antibody (XVIF9-G10)

            Advanced Verification
            45 Published Figures
            100 References
            View all (12) ATP1A3 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite ATP1A3 Monoclonal Antibody (XVIF9-G10)

            • Antibody Testing Data (9)
            • Published Figures (45)
            • Advanced Verification (1)
            Application
            ATP1A3 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 55

            ATP1A3 Antibody (MA3-915) in ICC/IF

            Immunofluorescence analysis of ATP1A3 was performed using SH-SY5Y cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with ATP1A3 Monoclonal Antibody (XVIF9-G10) (Product # MA3-915) at 5 µg/mL in 0.1% BSA, incubated at 4 degree celsius overnight and then labeled with Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor Plus 488 (Product # A32723), (1:2000), for ... View More
            View Product

            ATP1A3 Antibody in Immunocytochemistry (ICC/IF)
            ATP1A3 Antibody in Immunocytochemistry (ICC/IF)
            ATP1A3 Antibody in Immunocytochemistry (ICC/IF)
            ATP1A3 Antibody in Immunocytochemistry (ICC/IF)
            ATP1A3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ATP1A3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ATP1A3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Immunocytochemistry (ICC/IF)
            ATP1A3 Antibody in Immunocytochemistry (ICC/IF)
            ATP1A3 Antibody in Immunocytochemistry (ICC/IF)
            ATP1A3 Antibody in Immunocytochemistry (ICC/IF)
            ATP1A3 Antibody in Immunocytochemistry, Immunohistochemistry (ICC/IF, IHC)
            ATP1A3 Antibody in Immunocytochemistry (ICC/IF)
            ATP1A3 Antibody in Immunocytochemistry (ICC/IF)
            ATP1A3 Antibody in Immunohistochemistry (IHC)
            ATP1A3 Antibody in Immunohistochemistry (IHC)
            ATP1A3 Antibody in Immunohistochemistry (IHC)
            ATP1A3 Antibody in Immunohistochemistry (IHC)
            ATP1A3 Antibody in Western Blot, Immunoprecipitation (WB, IP)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody in Western Blot (WB)
            ATP1A3 Antibody

            Product Details

            MA3-915

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1 µg/mL
            View 65 publications 65 publications

            Immunohistochemistry (IHC)

            -
            View 19 publications 19 publications

            Immunohistochemistry (Paraffin) (IHC (P))

            1:50-1:200
            -

            Immunohistochemistry (Frozen) (IHC (F))

            3 µg/mL
            View 1 publication 1 publication

            Immunocytochemistry (ICC/IF)

            5 µg/mL
            View 12 publications 12 publications

            Flow Cytometry (Flow)

            1 µg / 10^6 cells
            -

            Immunoprecipitation (IP)

            -
            View 1 publication 1 publication

            Miscellaneous PubMed (Misc)

            -
            View 2 publications 2 publications
            Product Specifications

            Species Reactivity

            Bovine, Dog, Guinea pig, Human, Mouse, Non-human primate, Sheep, Rabbit, Rat

            Published species

            Amphibian, Bovine, Gerbil, Human, Mouse, Non-human primate, Pig, Rat, Shark

            Host/Isotype

            Mouse / IgG1

            Class

            Monoclonal

            Type

            Antibody

            Clone

            XVIF9-G10

            Immunogen

            Canine cardiac microsomes.

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1 mg/mL

            Purification

            Protein A

            Storage buffer

            0.02M potassium phosphate, pH 7.2, with 0.15M NaCl

            Contains

            0.05% sodium azide

            Storage conditions

            -20°C, Avoid Freeze/Thaw Cycles

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_2274447

            Product Specific Information

            MA3-915 detects sodium/potassium ATPase from human, monkey, bovine, sheep, canine, rabbit, guinea pig, mouse and rat tissue. This antibody is specific for the alpha-3 subunit.

            MA3-915 has been successfully used in Western blot and immunohistochemical procedures. By Western blot, this antibody detects an ~110 kDa protein representing the alpha-3 subunit of the sodium/potassium ATPase from canine skeletal muscle extract. Immunohistochemical staining of sodium/potassium ATPase in rat retina with MA3-915 yields a pattern consistent with plasma membrane localization.

            The MA3-915 antigen is canine cardiac microsomes.

            Target Information

            The sodium/potassium ATPase is an integral membrane enzyme found in all cells of higher organisms and is responsible for the ATP-dependent transport of sodium and potassium across the cell membrane. This membrane-bound enzyme is related to a number of other ATPases including sarcoplasmic and endoplasmic reticulum calcium ATPase (SERCA) and plasma membrane calcium ATPase (PMCA). The sodium/potassium ATPase consists of a large, multipass, transmembrane catalytic subunit, termed the alpha subunit, and an associated smaller glycoprotein, termed the beta subunit. Studies indicate that there are three isoforms of the alpha subunit (alpha 1, alpha 2, alpha 3) and two isoforms of the beta subunit (beta 1 and beta 2) encoded by two multigene families. The different isoforms of the sodium/potassium ATPase exhibit tissue-specific and developmental patterns of expression. The alpha 1 and beta mRNAs are present in all cell types examined, whereas the alpha 2 and alpha 3 mRNAs exhibit a more restricted pattern of cell-specific expression. The alpha-3 subunit has been found in neuronal and to skeletal and cardiac muscle, lung and stomach tissues.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: ATPase, Na+/K+ transporting, alpha 3 polypeptide; ATPase, Na+K+ transporting, alpha 3 subunit; EC 3.6.3.9; Na(+)/K(+) ATPase alpha(III) subunit; Na(+)/K(+) ATPase alpha-3 subunit; Na+, K+ activated adenosine triphosphatase alpha subunit; Na+/K+ -ATPase alpha 3 subunit; Na+/K+ ATPase 3; Sodium Potassium ATPase; Sodium pump 3; Sodium pump subunit alpha-3; sodium-potassium ATPase catalytic subunit alpha-3; sodium-potassium-ATPase, alpha 3 polypeptide; sodium/potassium-transporting ATPase alpha-3 chain; Sodium/potassium-transporting ATPase subunit alpha-3

            View more View less

            Gene Aliases: AHC2; ATP1A1; ATP1A3; Atpa-2; ATPa1a3; CAPOS; DYT12; RDP

            View more View less

            UniProt ID: (Human) P13637, (Rat) P06687, (Mouse) Q6PIC6

            View more View less

            Entrez Gene ID: (Sheep) 101103751, (Human) 478, (Bovine) 282598, (Dog) 612463, (Rabbit) 100349382, (Rat) 24213, (Guinea pig) 100712985, (Mouse) 232975

            View more View less

            Function(s)

            sodium:potassium-exchanging ATPase activity ATP binding D1 dopamine receptor binding heparan sulfate proteoglycan binding metal ion binding chaperone binding sodium:potassium-exchanging ATPase activity involved in regulation of cardiac muscle cell membrane potential steroid hormone binding protein binding cation-transporting ATPase activity nucleotide binding hydrolase activity primary active transporter

            Process(es)

            cellular sodium ion homeostasis memory adult locomotory behavior visual learning potassium ion import establishment or maintenance of transmembrane electrochemical gradient ATP hydrolysis coupled proton transport cellular potassium ion homeostasis ion transmembrane transport ionotropic glutamate receptor signaling pathway sodium ion export from cell response to drug cardiac muscle contraction cellular response to steroid hormone stimulus regulation of cardiac muscle cell membrane potential cell communication by electrical coupling involved in cardiac conduction response to glycoside regulation of cardiac conduction potassium ion transport sodium ion transport metabolic process transport ion transport sodium ion transmembrane transport potassium ion transmembrane transport
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-nml4b:80/100.66.75.22:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0