Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 12
This antibody is generated against the C-terminal region (665-1013aa) of ATP1A3 and detects the band around 100-113 kDa in SDS-PAGE.
Immunogen sequence: SEQIDEILQ NHTEIVFART SPQQKLIIVE GCQRQGAIVA VTGDGVNDSP ALKKADIGVA MGIAGSDVSK QAADMILLDD NFASIVTGVE EGRLIFDNLK KSIAYTLTSN IPEITPFLLF IMANIPLPLG TITILCIDLG TDMVPAISLA YEAAESDIMK RQPRNPRTDK LVNERLISMA YGQIGMIQAL GGFFSYFVIL AENGFLPGNL VGIRLNWDDR TVNDLEDSYG QQWTYEQRKV VEFTCHTAFF VSIVVVQWAD LIICKTRRNS VFQQGMKNKI LIFGLFEETA LAAFLSYCPG MDVALRMYPL KPSWWFCAFP YSFLIFVYDE IRKLILRRNP GGWVEKETYY (665-1013 aa encoded by BC015566)
The sodium/potassium ATPase is an integral membrane enzyme found in all cells of higher organisms and is responsible for the ATP-dependent transport of sodium and potassium across the cell membrane. This membrane-bound enzyme is related to a number of other ATPases including sarcoplasmic and endoplasmic reticulum calcium ATPase (SERCA) and plasma membrane calcium ATPase (PMCA). The sodium/potassium ATPase consists of a large, multipass, transmembrane catalytic subunit, termed the alpha subunit, and an associated smaller glycoprotein, termed the beta subunit. Studies indicate that there are three isoforms of the alpha subunit (alpha 1, alpha 2, alpha 3) and two isoforms of the beta subunit (beta 1 and beta 2) encoded by two multigene families. The different isoforms of the sodium/potassium ATPase exhibit tissue-specific and developmental patterns of expression. The alpha 1 and beta mRNAs are present in all cell types examined, whereas the alpha 2 and alpha 3 mRNAs exhibit a more restricted pattern of cell-specific expression. The alpha-3 subunit has been found in neuronal and to skeletal and cardiac muscle, lung and stomach tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase, Na+/K+ transporting, alpha 3 polypeptide; ATPase, Na+K+ transporting, alpha 3 subunit; EC 3.6.3.9; Na(+)/K(+) ATPase alpha(III) subunit; Na(+)/K(+) ATPase alpha-3 subunit; Na+, K+ activated adenosine triphosphatase alpha subunit; Na+/K+ -ATPase alpha 3 subunit; Na+/K+ ATPase 3; Sodium Potassium ATPase; Sodium pump 3; Sodium pump subunit alpha-3; sodium-potassium ATPase catalytic subunit alpha-3; sodium-potassium-ATPase, alpha 3 polypeptide; sodium/potassium-transporting ATPase alpha-3 chain; Sodium/potassium-transporting ATPase subunit alpha-3
Gene Aliases: AHC2; ATP1A1; ATP1A3; Atpa-2; Atpa1a3; CAPOS; DYT12; RDP
UniProt ID: (Human) P13637, (Rat) P06687, (Mouse) Q6PIC6
Entrez Gene ID: (Human) 478, (Rat) 24213, (Mouse) 232975
Molecular Function: primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support