Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: TDYFTAMAQE GWFPLLCVGL RAQWEDHHLQ DLQDSYGQ
Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase, H+/K+ exchanging, alpha polypeptide; ATPase, H+/K+ transporting, alpha polypeptide; Gastric H(+)/K(+) ATPase subunit alpha; gastric H+/K+ ATPase alpha subunit; gastric H,K-ATPase alpha subunit; gastric H,K-ATPase catalytic subunit; gastric hydrogen-potassium ATPase; H(+)-K(+)-ATPase alpha subunit; Potassium-transporting ATPase alpha chain 1; Proton pump
Gene Aliases: ATP4A; ATP6A
UniProt ID: (Human) P20648
Entrez Gene ID: (Human) 495
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support