Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IGRGQRELII GDRQTGKTSI AIDTIINQKR FNDGSDEKKK LYCIYVAIGQ KRSTVAQLVK RLTDADAMKY TIVVSATASD AAPLQYLAPY SGCSMGEYFR DNGKHALIIY DDLSKQAVAY RQMSLLLRRP PGREAYPGDV FYLHSRLLER AAKMNDAFGG GSLTALPVIE TQAGDVSAYI PTNVISITDG QIFLETELFY KGIRPAINVG LSVSRVGSAA QTRAMKQVAG TMKLELAQYR EVAAFAQFGS DLDAATQQLL SRGVRLTELL KQGQYSPMAI EEQVAVIYAG VRGYLDKLEP SKITKFENAF LSHVVSQHQA LLGTIRADGK ISEQSDAKLK EIVTNFLAGF EA
This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, using an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the alpha subunit of the catalytic core. Alternatively spliced transcript variants encoding the different isoforms have been identified. Pseudogenes of this gene are located on chromosomes 9, 2, and 16.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP synthase alpha chain, mitochondrial; ATP synthase F1 subunit alpha; ATP synthase subunit alpha, mitochondrial; ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit, isoform 1; ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit, isoform 1, cardiac muscle; ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit, isoform 2, non-cardiac muscle-like 2; ATP sythase (F1-ATPase) alpha subunit; epididymis secretory sperm binding protein Li 123m; iso; mitochondrial ATP synthetase, oligomycin-resistant; mitochondrial H+-ATP synthase alpha subunit; modifier of Min 2
Gene Aliases: AI035633; AL022851; AL023067; ATP5A; ATP5A1; ATP5AL2; ATP5F1A; ATPM; COXPD22; D18Ertd206e; hATP1; HEL-S-123m; MC5DN4; MOM2; OMR; ORM
UniProt ID: (Human) P25705, (Rat) P15999, (Mouse) Q03265
Entrez Gene ID: (Human) 498, (Rat) 65262, (Mouse) 11946
Molecular Function:
ATP synthase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support