FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat, Bovine and Orangutan
This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the gamma subunit of the catalytic core. Alternatively spliced transcript variants encoding different isoforms have been identified. This gene also has a pseudogene on chromosome 14.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP synthase F1 subunit gamma; ATP synthase gamma chain, mitochondrial; ATP synthase gamma-subunit gene encoding a mitochondrial protein; ATP synthase subunit gamma, mitochondrial; F-ATPase gamma subunit; F1 gamma; mitochondrial ATP synthase, gamma subunit 1
Gene Aliases: 1700094F02Rik; ATP5C; ATP5C1; ATP5CL1; ATP5F1C
UniProt ID: (Human) P36542, (Mouse) Q91VR2
Entrez Gene ID: (Human) 509, (Mouse) 11949
Molecular Function:
ATP synthase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support