Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLPAALLRR PGLGRLVRHA RAYAEAAAAP AAASGPNQMS FTFASPTQVF FNGANVRQVD VPTLTGAFGI LAAHVPTLQV LRPGLVVVHA EDGTTSKYFV SSGSIAVNAD SSVQLLAEEA VTLDMLDLGA AKANLEKAQA ELVGTADEAT RAEIQIRIEA NEALVKALE (1-168 aa encoded by BC002389 )
This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the delta subunit of the catalytic core. Alternatively spliced transcript variants encoding the same isoform have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP synthase F1 subunit delta; ATP synthase subunit delta, mitochondrial; F-ATPase delta subunit; mitochondrial ATP synthase complex delta-subunit precusor; mitochondrial ATP synthase, delta subunit
Gene Aliases: 0610008F14Rik; 1500000I11Rik; AA960090; AI876556; ATP5D; ATP5F1D; AU020773; C85518
UniProt ID: (Human) Q6FG90, (Mouse) Q9D3D9, (Rat) P35434
Entrez Gene ID: (Human) 513, (Mouse) 66043, (Rat) 245965
Molecular Function: ATP synthase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support