Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAPAVSGL SRQVRCFSTS VVRPFAKLVR PPVQVYGIEG RYATALYSAA SKQNKLEQVE KELLRVAQIL KEPKVAASVL NPYVKRSIKV KSLNDITAKE RFSPLTTNLI NLLAENGRLS NTQGVVSAFS TMMSVHRGEV PCTVTSASPL EEATLSELKT VLKSFLSQGQ VLKLEAKTDP SILGGMIVRI GEKYVDMSVK TKIQKLGRAM REIV (1-213 aa encoded by BC021233)
The protein encoded by this gene is a component of the F-type ATPase found in the mitochondrial matrix. F-type ATPases are composed of a catalytic core and a membrane proton channel. The encoded protein appears to be part of the connector linking these two components and may be involved in transmission of conformational changes or proton conductance.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP synthase peripheral stalk subunit OSCP; ATP synthase subunit O, mitochondrial; ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit (oligomycin sensitivity conferring protein); ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit; DNA segment, Chr 12, Wayne State University 28, expressed; human ATP synthase OSCP subunit; Oligomycin sensitivity conferral protein; oligomycin sensitivity conferral protein oscp-like protein; oligomycin sensitivity conferring protein; OSCP; Sperm flagella protein 4
Gene Aliases: ATP5O; ATP5PO; ATPO; D12Wsu28e; HMC08D05; OSCP
UniProt ID: (Human) P48047, (Mouse) Q9DB20, (Rat) Q06647
Entrez Gene ID: (Human) 539, (Mouse) 28080, (Rat) 192241
Molecular Function: ATP synthase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support