Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Vacuolar-type H+-ATPase (V-ATPase) is a multisubunit enzyme responsible for acidification of eukaryotic intracellular organelles. V-ATPases pump protons against an electrochemical gradient, thereby synthesizing ATP. A peripheral V1 domain, which is responsible for ATP hydrolysis, and an integral V0 domain, which is responsible for proton translocation, compose the V-ATPase. Nine subunits (A-H) make up the V1 domain and five subunits (a, d, c, c' and c"") make up the V0 domain. ATP6AP1 (ATPase, H+ transporting, lysosomal accessory protein 1), also known as 16A, CF2, Ac45, XAP3, ATP6S1, VATPS1 (vacuolar ATP synthase S1 accessory protein) or ATP6IP1, is a type I transmembrane, V-ATPase accessory protein that is predominantly expressed in endocrine and neuronal cells. ATP6AP1 is responsible for targeting the V-ATPase enzyme to specialized complex vacuolar systems. Via its cytoplasmic tail, ATP6AP1 interacts with subunits of the V0 domain. The disruption of this interaction in osteoclasts results in impaired bone resorption, suggesting an important role for ATP6AP1 in proper osteoclastic bone resorption.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase, H+ transporting, lysosomal (vacuolar proton pump), subunit 1; ATPase, H+ transporting, lysosomal accessory protein 1; ATPase, H+ transporting, lysosomal interacting protein 1; H-ATPase subunit; MGC129781; Protein XAP-3; V-ATPase Ac45 subunit; V-ATPase S1 accessory protein; V-ATPase subunit S1; V-type proton ATPase subunit S1; Vacuolar proton pump subunit S1
Gene Aliases: 16A; Ac45; ATP6AP1; ATP6IP1; ATP6S1; CF2; VATPS1; XAP-3; XAP3
UniProt ID: (Human) Q15904
Entrez Gene ID: (Human) 537
Molecular Function:
ATP synthase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support