Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTGLALLYSG VFVAFWACAL AVGVCYTIFD LGFRFDVAWF LTETSPFMWS NLGIGLAISL SVVGAAWGIY ITGSSIIGGG VKAPRIKTKN LVSIIFCEAV AIYGIIMAIV ISNMAEPFSA TDPKAIGHRN YHAGYSMFGA GLTVGLSNLF CGVCVGIVGS GAALADAQNP SLFVKILIVE IFGSAIGLFG VIVAILQTSR VKMGD
This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c'', and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is part of the transmembrane V0 domain and is the human counterpart of yeast VMA16. Two alternatively spliced transcript variants that encode different proteins have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase, H+ transporting, lysosomal 21kDa, V0 subunit b; ATPase, H+ transporting, lysosomal, 21-KD, V0 subunit C-prime, prime; ATPase, H+ transporting, lysosomal, subunit F; H(+)-transporting two-sector ATPase, subunit F; hATPL; V-ATPase 21 kDa proteolipid subunit; V-ATPase 21 kDa proteolipid subunit c''; V-ATPase subunit c''; V-type proton ATPase 21 kDa proteolipid subunit c''; vacuol; vacuolar ATP synthase 21 kDa proteolipid subunit; vacuolar proton pump 21 kDa proteolipid subunit; Vacuolar proton pump 21 kDa proteolipid subunit c''
Gene Aliases: ATP6F; ATP6V0B; HATPL; VMA16
UniProt ID: (Human) Q99437
Entrez Gene ID: (Human) 533
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support