Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
This antibody is predicted to react with bovine, canine, equine and rat based on sequence homology.
ATP6V0C is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c', and d. ATP6V0C encodes the V0 subunit c.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase, H+ transporting, lysosomal (vacuolar proton pump) 16 kDa; ATPase, H+ transporting, lysosomal 16kD, V0 subunit C; ATPase, H+ transporting, lysosomal 16kDa, V0 subunit c; ATPase, H+ transporting, lysosomal 2; H(+)-ATPase (mvp); H(+)-transporting two-sector ATPase, 16 kDa subunit; PL16; V-ATPase 16 kDa proteolipid subunit; V-ATPase 16 kDa proteolipid subunit c; V-type proton ATPase 16 kDa proteolipid subunit; V-type proton ATPase 16 kDa proteolipid subunit c; vacuolar ATP synthase 16 kDa proteolipid subunit; vacuolar H+ ATPase proton channel subunit; vacuolar proton pump 16 kDa proteolipid subunit; Vacuolar proton pump 16 kDa proteolipid subunit c
Gene Aliases: ATP6C; Atp6c2; ATP6L; ATP6V0C; ATPL; Atpl-rs1; Mvp; PL16; VATL; Vma3; VPPC
UniProt ID: (Human) P27449, (Mouse) P63082
Entrez Gene ID: (Human) 527, (Mouse) 11984
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support