Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAMEIDSRPG GLPGSSCNLG AAREHMQAVT RNYITHPRVT YRTVCSVNGP LVVLDRVKFA QYAEIVHFTL PDGTQRSGQV LEVAGTKAIV QVFEGTSGID ARKTTCEFTG DILRTPVSED MLGRVFNGSG KPIDKGPVVM AEDFLDINGQ PINPHSRIYP EEMIQTGISP IDVMNSIARG QKIPIFSAAG LPHNEIAAQI CRQAGLVKKS KAVLDYHDDN FAIVFAAMGV NMETARFFKS DFEQNGTMGN VCLFLNLAND PTIERIITPR LALTTAEFLA YQCEKHVLVI LTDMSSYAEA LREVSAAREE VPGRRGFPGY MYTDLATIYE RAGRVEGRGG SITQIPILTM PNDDITHPIP DLTGFITEGQ IYVDRQLHNR QIYPPINVLP SLSRLMKSAI GEGMTRKDHG DVSNQLYACY AIGKDVQAMK AVVGEEALTS EDLLYLEFLQ KFEKNFINQG PYENRSVFES LDLGWKLLRI FPKEMLKRIP QAVIDEFYSR EGALQDLAPD TAL
Vacuolar-type H+-ATPase (V-ATPase) is a multisubunit enzyme responsible for acidification of eukaryotic intracellular organelles. V-ATPases pump protons against an electrochemical gradient, while F-ATPases reverse the process, thereby synthesizing ATP. A peripheral V1 domain, which is responsible for ATP hydrolysis, and a integral V0 domain, which is responsible for proton translocation, compose V-ATPase. Nine subunits (A-H) make up the V1 domain and five subunits (a, d, c, c' and c") make up the V0 domain. Like F-ATPase, V-ATPase most likely operates through a rotary mechanism. The V-ATPase V1 B subunit exists as two isoforms. In the inner ear, the V-ATPase B1 isoform functions in proton secretion and is required to maintain proper endolymph pH and normal auditory function. The gene encoding the human V-ATPase B1 isoform maps to chromosome 2cen-q13. Mutations in this gene cause distal renal tubular acidosis associated with sensorineural deafness. The V-ATPase B2 isoform is expressed in kidney and is the only B isoform expressed in osteoclasts.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1; Endomembrane proton pump 58 kDa subunit; H(+)-transporting two-sector ATPase, 58kD subunit; H+-ATPase beta 1 subunit; V-ATPase B1; V-ATPase B1 subunit; V-ATPase subunit B 1; V-type proton ATPase subunit B, kidney isoform; vacuolar proton pump 3; Vacuolar proton pump subunit B 1; vacuolar proton pump, subunit 3
Gene Aliases: ATP6B1; ATP6V1B1; RTA1B; VATB; VMA2; VPP3
UniProt ID: (Human) P15313
Entrez Gene ID: (Human) 525
Molecular Function: ATP synthase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support