Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
ATP6V1C2 encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A, three B, and two G subunits, as well as a C, D, E, F, and H subunit. The V1 domain contains the ATP catalytic site. This gene encodes alternate transcriptional splice variants, encoding different V1 domain C subunit isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C2; V-ATPase C2 subunit; V-ATPase subunit C 2; V-type proton ATPase subunit C 2; Vacuolar proton pump subunit C 2
Gene Aliases: ATP6C2; ATP6V1C2; VMA5
UniProt ID: (Human) Q8NEY4
Entrez Gene ID: (Human) 245973
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support