Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAGRGKLIA VIGDEDTVTG FLLGGIGELN KNRHPNFLVV EKDTTINEIE DTFRQFLNRD DIGIILINQY IAEMVRHALD AHQQSIPAVL EIPSKEHPYD AAKDSILRRA RGMFTAEDLR (1-119 aa encoded by BC107854)
ATP6V1F encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c", and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is the V1 domain F subunit protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adenosinetriphosphatase 14k chain; ATPase, H+ transporting, lysosomal 14kDa, V1 subunit F; ATPase, H+ transporting, V1 subunit F; ATPase, vacuolar, 14 kD; H(+)-transporting two-sector ATPase, 14kD subunit; V-ATPase 14 kDa subunit; V-ATPase F subunit; V-ATPase subunit F; V-type proton ATPase subunit F; vacuolar ATP synthase subunit F; vacuolar proton pump F subunit; Vacuolar proton pump subunit F
Gene Aliases: 1110004G16Rik; ATP6S14; ATP6V1F; VATF; Vma7
UniProt ID: (Human) Q16864, (Mouse) Q9D1K2
Entrez Gene ID: (Human) 9296, (Mouse) 66144
Molecular Function: ATP synthase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support