Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MGPSIPAKTR EVLVSHLASY NTWALQGIEF VAAQLKSMVL TLGLIDLRLT VEQAVLLSRL EEEYQIQKWG NIEWAHDYEL QELRARTAAG TLFIHLCSES TTVKHKLLKE
This gene encodes an assembly factor for the F(1) component of the mitochondrial ATP synthase. This protein binds specifically to the F1 alpha subunit and is thought to prevent this subunit from forming nonproductive homooligomers during enzyme assembly. This gene is located within the Smith-Magenis syndrome region on chromosome 17. An alternatively spliced transcript variant has been described, but its biological validity has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP synthase mitochondrial F1 complex assembly factor 2; ATP12 homolog
Gene Aliases: ATP12; ATP12p; ATPAF2; LP3663; MC5DN1
UniProt ID: (Human) Q8N5M1
Entrez Gene ID: (Human) 91647
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support