Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: APRGSRREPG GWGAGASAPV EDDSDAETYG EENDEQGNYS KRKIVSNWDR YQDIEKEVNN ESGESQRGTD FSVLLSSAGD SFSQFRFAEE KEWDSEASCP
Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer. Aven is a recently discovered protein that blocks apoptosis induced by Apaf-1 and caspase-9. It is thought that Aven functions by binding to Bcl-xL, an antiapoptotic member of the Bcl-2 family, and to Apaf-1, possibly interfering with the ability of Apaf-1 to self-associate, suggesting that Aven impedes Apaf-1-mediated caspase activation. Higher levels of Aven mRNA are seen in patients with acute leukemia than in control patients, suggesting that Aven may be useful as a prognostic indicator in leukemia patients.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: apoptosis, caspase activation inhibitor; Cell death regulator Aven; programmed cell death 12
Gene Aliases: 1700013A01Rik; 1700056A21Rik; AVEN; mAven-L; mAven-S; PDCD12
UniProt ID: (Human) Q9NQS1, (Mouse) Q9D9K3
Entrez Gene ID: (Human) 57099, (Rat) 311299, (Mouse) 74268
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support