Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MDALVEDDIC ILNHEKAHKR DTVTPVSIYS GDESVASHFA LVTAYEDIKK RLKDSEKENS LLKKRIRFLE EKLIARFEEE TSSVGREQVN KAYHAYREVC IDRDNLKSKL DKMNKDNSES LKVLNEQLQS KEVELLQLRT EVETQQVMRN LNPPSSNWEV EKLSCDLKIH GLEQELELMR KECSDLKIEL QKAKQTDPYQ EDNLKSRDLQ KLSISSDNMQ HAYWELKREM SNLHLVTQVQ AELLRKLKTS TAIKKGKSLF TGNH
AZI2, or NAP1, contributes to the activation of NFKB (Fujita et al., 2003 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5-azacytidine induced gene 2; 5-azacytidine-induced protein 2; NAK-associated protein 1; NF-kappa-B-activating kinase-associated protein 1; TILP
Gene Aliases: AZ2; AZI2; NAP1; TBKBP2; TILP
UniProt ID: (Human) Q9H6S1
Entrez Gene ID: (Human) 64343
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support