Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
Anti-Abi1 is directed against mouse Abi1. A BLAST analysis was used to suggest cross reactivity with this protein in human and rat based on 100% homology for the immunogen sequence. Reactivity against homologues from other sources is not known.
This gene encodes a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. The nucleolar protein encoded by this gene interacts in a hormone-dependent manner with nuclear receptors, and represses their transcriptional activity. Alternative splice variants that encode different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Abelson interactor 1; Abi-1; Abl interactor 1; Ablphilin-1; e3B1; EL2; eps8 binding protein; Eps8 SH3 domain-binding protein; Eps8-binding protein; HS3; spectrin SH3 domain binding protein 1; Spectrin SH3 domain-binding protein 1; SPH3; SPTA
Gene Aliases: Abi1; E3B1; NAP1; Ssh3bp1
UniProt ID: (Mouse) Q8CBW3
Entrez Gene ID: (Mouse) 11308
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support