Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: ACHAAMRPP QCLLHTPSLA SPLLLLLVWL LGGGVGAEGR EDAELLVTVR GGRLRGIRLK TPGGPVSAFL GIPFAEPPMG PRRFLPPEPK QPWSGVVDAT TFQSVCYQYV DTLYPGFEGT EMWNPNRELS EDCLYLNVWT PYPRPTSPTP VIVWIYGGGF YSGASSLDVY DGRFLVQAER TVLVSMNYRV GAFGFLALPG SREAPGNVGL LDQRLALQWV QENVAAFGGD PTSVTLFGES AGAASVGMHL LSPPSRGLFH RAVLQSGAPN GPWATVGMGE ARRRATQLAH LVGCPPGGTG GNDTELVACL RTRPAQVLVN HEWHVLPQES VFRFSFVPVV DGDFLSD (1-351 aa encoded by BC094752)
Acetylcholinesterase hydrolyzes the neurotransmitter, acetylcholine at neuromuscular junctions and brain cholinergic synapses, and thus terminates signal transmission. The Protein is also found on the red blood cell membranes, where it constitutes the Yt blood group antigen. Acetylcholinesterase exists in multiple molecular forms which possess similar catalytic properties, but differ in their oligomeric assembly and mode of cell attachment to the cell surface. The major form of acetylcholinesterase found in brain, muscle and other tissues is the hydrophilic species, which forms disulfide-linked oligomers with collagenous, or lipid-containing structural subunits.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acetylcholinesterase; acetylcholinesterase (Yt blood group); AChE; apoptosis-related acetylcholinesterase; EC 3.1.1.7; glycolipid-anchored form of acetylcholinesterase; OTTHUMP00000211349; OTTHUMP00000211356; Yt blood group
Gene Aliases: ACEE; ACHE; ARACHE; N-ACHE; YT
UniProt ID: (Human) Q9BXP7, (Mouse) P21836, (Rat) P37136
Entrez Gene ID: (Human) 43, (Mouse) 11423, (Rat) 83817
Molecular Function: esterase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support